Resistance Blocks
200 Ohm Resistance block
Catalog # 470121-100
Supplier: Wards
Supplier #: PH1143H
Jump to:
Product Details & Documents
Economical yet Sturdy
- Simple to use
- Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Spécifications
- Désignation:200 Ohm Resistance block