Resistance Blocks
Resistance Blocks
200 Ohm Resistance block
Catalog # 470121-100
Supplier:   Wards
Supplier #:  PH1143H
Resistance Blocks
Catalog # 470121-100
Supplier:  Wards
Resistance Blocks
Resistance Blocks
200 Ohm Resistance block
Liquidation
$3.10
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Economical yet Sturdy
  • Simple to use
  • Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Spécifications
  • Désignation:
    200 Ohm Resistance block