Resistance Blocks
10 Ohm Resistance block
  470121-092
 :   Wards
 :  PH1143D
Resistance Blocks
  470121-092
 :  Wards
Resistance Blocks
Resistance Blocks
10 Ohm Resistance block
$3.10
  31/12/2026
Each
Economical yet Sturdy
  • Simple to use
  • Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
  • Désignation:
    10 Ohm Resistance block