Resistance Blocks
Resistance Blocks
5 Ohm Resistance block
Catalog # 470121-090
Supplier:   Wards
Supplier #:  PH1143C
Resistance Blocks
Catalog # 470121-090
Supplier:  Wards
Resistance Blocks
Resistance Blocks
5 Ohm Resistance block
Price
9,95 $
Log in for availability
Each
Product Details & Documents
Economical yet Sturdy
  • Simple to use
  • Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
Specifications
  • Désignation:
    5 Ohm Resistance block