Resistance Blocks
1 Ohm Resistance block
470121-086
: Wards
: PH1143A
Economical yet Sturdy
- Simple to use
- Safe to handle
Students can use these resistance units to construct various types of circuits. Built with a high stability metal film element, these durable resistance units are more economical that resistance coils. Accuracy: 1%. 1W.
- Désignation:1 Ohm Resistance block