Edvotek® Variable Micropipettes | Avantor
Edvotek® Variable Micropipettes
Pipette Carousel Stand for Six Pipets, Light Gray
Catalog # 470121-052
Supplier:   Edvotek
Fournisseur #:  796
Edvotek® Variable Micropipettes
Catalog # 470121-052
Supplier:  Edvotek
Edvotek® Variable Micropipettes
Edvotek® Variable Micropipettes
Pipette Carousel Stand for Six Pipets, Light Gray
Price
$101.63
Log in pour la disponibilité
Each
Product Details & Documents
Our newly re-designed variable micropipettes are sturdily designed with volumes ranging from 0.1 to 5000 µl.
  • New and improved
  • High quality, same price
  • More durable and ergonomic
  • Lifetime warranty
  • Light gray color
They are easy to use, highly accurate and use standard micropipette tips. The volume is easily selected by twisting the top. The lightweight design and tip ejector make operation fast and easy.
Ordering information: A tool and instructions are included for self-calibration.
Spécifications
  • Désignation:
    Pipette Carousel Stand for Six Pipets, Light Gray