Mechanical Engineering Models | Avantor
Mechanical Engineering Models
Worm Gear
Catalog # 470120-720
Supplier:   Wards
Fournisseur #:  PH0366-TM-6
Mechanical Engineering Models
Catalog # 470120-720
Supplier:  Wards
Mechanical Engineering Models
Mechanical Engineering Models
Worm Gear
Price
$77.60
Log in pour la disponibilité
Each
Product Details & Documents
Clearly Observe Inner Workings
  • Demonstrate the most fundamental and most frequently used mechanisms with these mechanical engineering models
  • All components are precisely designed and can be assembled with clear sides to facilitate teaching
Useful for introducing engineering concepts, with models that are easy to assemble and disassemble. These models are great to visually demonstrate how the internal mechanisms of car transmissions, snowmobiles, elevators, and other mechanical items work. Students will be able to see things from how a simple transmission works with three forward gears and one reverse to the typical cone clutch drive used in many snowmobiles.
Spécifications
  • Désignation:
    Worm Gear