VWR® Volumetric Flasks, Class B | Avantor
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 100, Tolerance, ±mL: 0.16
Catalog # 470120-670
Supplier:   VWR International
VWR® Volumetric Flasks, Class B
Catalog # 470120-670
Supplier:  VWR International
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 100, Tolerance, ±mL: 0.16
Price
$583.35
Log in pour la disponibilité
Pack of 12
Product Details & Documents
Our new high-quality borosilicate volumetric flasks meet ASTM standards and are a great value.
  • Borosilicate glass construction
  • Snap-on caps
  • Graduated line for accurate measurements
Class B with capacity tolerances twice those specified for Class A volumetric work. The necks are tooled to fit the included snap-caps. With graduation line for accurate measurements.
Spécifications
  • Désignation:
    VWR® Volumetric Flasks — Class B, Cap., mL: 100, Tolerance, ±mL: 0.16