Child CPR Training Torso | Avantor
Child CPR Training Torso
Carrying Bag
Catalog # 470119-760
Supplier:   Nasco
Fournisseur #:  100-2252
Child CPR Training Torso
Catalog # 470119-760
Supplier:  Nasco
Child CPR Training Torso
Child CPR Training Torso
Carrying Bag
Price
$240.85
Log in pour la disponibilité
Each
Product Details & Documents
Convenient And Economical Child Cpr Practice
  • Realistic chest rise
  • Anatomical landmarks such as sternum, ribcage and sub-sternal notch
  • 3 airway/lung/face shields included
This economical CPR torso uses a single-use airway/lung/face shield system which requires no cleaning, no disinfecting and no disassembly. Features include: Realistic chest rise, Anatomical landmarks such as sternum, ribcage and sub-sternal notch, 3 airway/lung/face shields
Spécifications
  • Désignation:
    Carrying Bag
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.