I.V. Training Hand
IV Hand
Catalog # 470119-424
Supplier: American 3B Scientific
Supplier #: 1005754
Jump to:
Product Details & Documents
Basic Hand Injection Practice
- Basic I.V. training introduction
- Supple skin allows for vein palpation
This training aid was designed for practice in giving IVs. It permits detailed exercises in venipuncture so that the technique can be mastered on a realistic model before actually administering to patients. The students can palpate the hand, which is made of supple, resilient plastic with a human feel. The vein is located, the needle introduced, and blood is withdrawn or fluid injected. The model can be used repeatedly. Complete with blood powder and carrying tray.
Specifications
- Désignation:IV Hand