Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Dehydrated Media

Dehydrated Media Tryptic Soy Agar, 114 g, Bag
Catalog # 470113-158
 :  LD100A
Dehydrated Media
Catalog # 470113-158
CAS Number:  
Dehydrated Media

Dehydrated Media Tryptic Soy Agar, 114 g, Bag

CAS Number:  
Price
48,60 $
Log in pour la disponibilité
Each
Product Details & Documents

Dehydrated Media is suitable for almost all educational applications.

  • For Use with Plates, Slants, and Broth Media
  • Instructions Included
  • Sealed Bottle
  • Expiration Date on Bottle

It arrives in a sealed bottle with an expiration date and complete instructions for preparation. Once items have shipped, they have three or more years before expiration.

Agar and components are used for plates or slants. Assuming a 20 mL fill for a 100 x 15 mm petri dish, each 114 g bottle will fill at least 100 dishes; each 454 g bottle will fill at least 400. Broth media are used for tube cultures. Assuming a 12 mL fill for a 20 x 75 mm tube, each 114 g bottle will fill at least 1000 tubes; each 454 g bottle will fill at least 2000.

Spécifications

Spécifications

  • Type de conditionnement:
    Bag
  • Désignation:
    Dehydrated Media Tryptic Soy Agar
  • Taille:
    114 g

Spécifications