Heat Transfer Activity
Replacement Insulated Containers, —
470112-184
Apparatus Shows Effect of Heated Water on Other Materials
- Literature included
- Observe transfer of heat under different conditions
Heat energy cannot be seen, but with this calorimeter featuring an aluminum transfer bar, students can observe heat transfer under a variety of different conditions by measuring the temperatures of the insulated containers before and after connecting them with the metal bar. You can also vary the experiment by using water of different temperatures, or unequal amounts of liquid.
: Please note, lids are not included with replacement containers. Materials included are Calorimeters, Aluminum transfer bar, thermometers. Materials Required but not Included: Boiling water, Wood Alcohol, 100 mL Graduated cylinder, Timer/stopwatch.
- Désignation:Replacement Insulated Containers
- No. of Lab Groups:—