Plane Mirrors | Avantor
Plane Mirrors
Plexiglas mirrors, front surface silvered, 75x125x3 mm
Catalog # 470111-294
Supplier:   Piedmont Plastics
Fournisseur #:  3512P
Plane Mirrors
Catalog # 470111-294
Supplier:  Piedmont Plastics
Plane Mirrors
Plane Mirrors
Plexiglas mirrors, front surface silvered, 75x125x3 mm
Price
$48.65
Log in pour la disponibilité
Pack of 6
Product Details & Documents
All Types and Sizes for All Optical Applications
  • Front surface silvering eliminates double reflections
  • All produce virtual images
These plane mirrors are rectangular. They are available in glass or shatterproof Plexiglas. Back surface silvered mirrors are less expensive, and ideal for general use. The silvered back surface is painted for durability. Front surface silvering eliminates double reflections at the unsilvered surface, giving a brighter, sharper image. However, these mirrors are more easily damaged by scratching.
Spécifications
  • Désignation:
    Plexiglas mirrors, front surface silvered, 75x125x3 mm