Equilateral Prisms
Equilateral Prisms, Glass, 25 x 75 mm
Catalog # 470111-280
Supplier: Wards
Fournisseur #: PH0554E
Jump to:
Product Details & Documents
For all Optical Applications
- These prisms are polished on all faces
- They are made of shatter-resistant acrylic material or optical quality glass
These prisms are polished on all faces. They are made of shatter-resistant acrylic material or optical quality glass. The length of the sides and the height are given for each prism.
Spécifications
- Désignation:Equilateral Prisms, Glass, 25 x 75 mm