Rudiger® Small Scale Human Skeleton Models | Avantor
Rudiger® Small Scale Human Skeleton Models
Small Scale Human Skeletons, With Painted Muscles
Catalog # 470104-878
Fournisseur #:  MI200.1
Rudiger® Small Scale Human Skeleton Models
Catalog # 470104-878
Rudiger® Small Scale Human Skeleton Models
Rudiger® Small Scale Human Skeleton Models
Small Scale Human Skeletons, With Painted Muscles
Price
$953.85
Log in pour la disponibilité
Each
Product Details & Documents
These Small Scale Skeletal Models Are Extremely Accurate.
  • Desktop Sized Skeletal Preparations For Easy Study
  • Bones Individually Cast, Allowing For Maximum Articulation
  • 5 Year Manufacturers Warranty
Both models feature high-quality bone definition, flexible joint structures, and a dissectible skull with a spring-held jaw to expose interior detail such as formamina, brain areas, and sinus channels. The model with painted muscles also presents 174 muscle origins and insertions, numbered and color-coded in red (origins) and blue (insertions) on one side only for comparison purposes. A key, which is included, identifies each of the muscles. Both models come fully assembled and include an easily assembled (no tools required) tubular mounting stand on a wood base.
Spécifications
  • Désignation:
    Small Scale Human Skeletons, With Painted Muscles