Rapitest Soil Test Kit | Avantor
Rapitest Soil Test Kit
Test kit, soil pH, Rapitest, 10
Catalog # 470104-846
Supplier:   Luster Leaf
Fournisseur #:  1612
Rapitest Soil Test Kit
Catalog # 470104-846
Supplier:  Luster Leaf
CAS Number:  
Rapitest Soil Test Kit
Rapitest Soil Test Kit
Test kit, soil pH, Rapitest, 10
CAS Number:  
Price
$8.30
Log in pour la disponibilité
Each Case of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perform qualitative soil tests without the use of chemical reagents and test tubes.
  • No hazardous compounds required for testing
  • Colorimetric determination of soil components
  • Instructions included
This kit lets students use a simple three-step process involving water, tablets and color comparators. To test soil, simply mix it with water and let it settle. Then pour the liquid off into the color comparator, add a tablet and compare the liquid with the color film chart for a test reading.
470104-846 includes 10 pH soil test kits and fully instructions.
470175-090 includes 10 tests each for pH, nitrogen, phosphorus and potash and full instructions.
Spécifications
Spécifications
  • Désignation:
    Test kit, soil pH, Rapitest
  • No. of Tests:
    10