Replacement Bulbs
Replacement Bulb, with Bayonet Base, 110 V, 15 W
Catalog # 470103-538
Supplier: VWR
Fournisseur #: 1101002400482
Jump to:
Product Details & Documents
Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.
Spécifications
- Désignation:Replacement Bulb, with Bayonet Base
- Electrical:110 V, 15 W