LaMotte® Secchi Disk
Secchi Disk Only, 8"
470101-430
: LaMotte
: 0171
This 8", black and white disk is used to determine the turbidity of natural water.
- Black and White design
- Sounding lead and line included
- Function: to determine the turbidity of water
- Aligned with GLOBE protocols
The disk is raised or lowered several times to determine the average depth at which it disappears from sight.
: 470112-626 is complete set, containing a secchi disk and a calibrate line. 470004-364 is the standalone rope, and 470101-430 the standalone disk.
- Désignation:Secchi Disk Only
- O.D.:8"