Sandpaper
Medium grit, 2×6"
Catalog # 470040-328
Supplier: POWER TOOLS & ABRASIVES, INC.
Fournisseur #: 2X6AMED
Jump to:
Product Details & Documents
Medium grade, 180 grit. 9×11" sheets.
Spécifications
- Désignation:Medium grit
- Dimensions:2×6"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



