Basic Power Supply Accessory Kit
Basic power supply accessory kit
Catalog # 470039-456
Supplier: CAL TEST ELECTRONICS SE
Supplier #: CT3374A
Jump to:
Product Details & Documents
Useful tools for any power supply.
This basic kit provides the necessary test leads and alligator clips required when starting to use your general-purpose bench-top AC or DC power supply. The kit features leads with high capacity silicone jacketed wire with retractile sleeve to straight fixed sheathed banana plugs. Two insulated alligator clips make the final connections. The four piece kit comes package in a clear PVC pouch for convenient storage. RoHS compliant.
Spécifications
- Désignation:Basic power supply accessory kit