Solid Balls for Physics Experiments
Solid Steel Ball, 13 mm, ½"
470032-360
: PNBS13-S
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
- Désignation:Solid Steel Ball, 13 mm
- Diameter:½"