Basic Buddy® Budget CPR Torsos
Budget CPR Torso
Catalog # 470029-890
Supplier: Nasco
Fournisseur #: LF03693
Jump to:
Product Details & Documents
Buy Just Enough For Your Needs.
- Includes replaceable lung bags
- Mouth protection is included on lung bags for sanitary use
- Available as individual, or bundles of trainers
Inexpensive manikins designed for teaching individuals or large groups the life-saving techniques of CPR. The one-piece, disposable lung/mouth protection bags make this manikin completely sanitary. The airway opens using the head tilt/chin lift method and there is a visible chest rise when ventilated. The xiphoid process provides an anatomical reference point for hand placement and compressions. The manikin also features both adult and child capabilities.
Each trainer comes with 10 lung mouth protection bags. Bundles are also supplied with 1 carrying / storage bag for every 5 Units.
Spécifications
- Désignation:Budget CPR Torso