Forensic Science Wound Simulation Kit
Product Details & Documents
Basic Trauma Evidence Gathering Unit
- From simple puncture wounds associated with sharp instrument trauma to firearms entry and exit wounds, this kit is critical in studying forensics
Forensic Science Wound Simulation Kit
From simple puncture wounds associated with sharp instrument trauma to firearms entry and exit wounds, this kit is critical in studying forensics. Especially useful in crime scene simulations, the wound set asks students: Can you recognize pattern ligature in strangulation? What was the murder weapon? What does a single-bladed knife stab wound look like compared to a double-edged blade wound? What are the differences between the wound of mechanism for suicide and hesitation marks? Each of these questions and more are addressed with a set of simulated wounds that also includes moulage wax, coagulant blood, and a selection of makeup for designing your own wounds of bruising, blanching, and lividity.
The entire kit comes placed in a sturdy carry case for protection and ease of transportation to the training or simulation site. This innovative kit contains carefully sculpted models of injuries taken from actual victim’s wounds and reviewed by forensic education professionals insuring real scene representations of trauma commonly found on violent crime victims. The details in the wounds are designed to determine type of weapon, direction of force, and other related details that only come with realistic trauma reproduction.
Wounds include: Petechiae; Ecchymosis; Pattern Ligature; Slash; Shotgun close range; Shotgun; Intermediate range; Intermediate range round; Intermediate range flake; Exit gunshot; Contact gunshot contact handgun; Contact wound; Screwdriver; Bite partial; Bite complete; Hesitation; Single-edge knife; Double-edge knife; Blunt bat; Bunny ears exit wound; Exit wound ice pick; Compound fracture humerus; Foreign body protrusion; Large laceration; Medium laceration large compound fracture; and 2nd degree burn.
Size of case: 21" x 13" x 15".
Forensic Science Wound Package
Special stick-on wounds add to the realism of simulated crime scenes and activities, while teaching an important aspect of forensic investigation. Supplement the simulated wounds in the kit with these stickers. The eighteen sticker wounds include Petechiae; Ecchymosis; Pattern Ligature; Slash; Shotgun close range; Shotgun; Intermediate range; Intermediate range round; Intermediate range flake; Exit gunshot; Contact gunshot contact handgun; Contact wound; Screwdriver; Bite partial; Bite complete; Hesitation; Single-edge knife; Double-edge knife; Blunt bat; Bunny ears exit wound; Exit wound ice pick.
Spécifications
- Désignation:Forensic Science Wound Simulation Kit