Viruses Microslide, Set | Avantor
Viruses Microslide, Set
Microslides, The Virus set, set of 15
Catalog # 470022-148
Fournisseur #:  T-97-15B
Viruses Microslide, Set
Catalog # 470022-148
Viruses Microslide, Set
Viruses Microslide, Set
Microslides, The Virus set, set of 15
Price
$291.65
Log in pour la disponibilité
Box of 15
Product Details & Documents
Delve into the world of microbes and viruses.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
How small is a virus? Do viruses look like our cells or even like other viruses? Compare and contrast disease-causing and helpful viruses.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Lambda phage on E. coli (em, 45,000X)
Disrupted phage (em, 280,000X)
Human viruses:
Influenza virus (em, 170,000X) and Polio virus (em, 290,000X)
Measles virus attack (fs, 400)
Animal viruses:
Newcastle disease (em, 20,000X) and Vaccinia virus (em, 120,000X)
Viruses emerging from cell (em, 90,000X)
Cancer viruses:
Papilloma virus (em, 7,000X) and Rauscher leukemia virus (em, 90,000X)
Box of 15 Sets also Includes:
Background subject information
Reproducible student worksheets with answer keys
Spécifications
  • Désignation:
    Microslides, The Virus set, set of 15