Cell Structure Microslides | Avantor
Cell Structure Microslides
Microslide, Cell Structure, Set of 15 (Box)
Catalog # 470022-104
Fournisseur #:  T-58-15B
Cell Structure Microslides
Catalog # 470022-104
Cell Structure Microslides
Cell Structure Microslides
Microslide, Cell Structure, Set of 15 (Box)
Price
$291.85
Log in pour la disponibilité
Box of 15
Product Details & Documents
Explore organelles up close. Each Microslide includes a slide featuring 8 related 35mm images photographed through a microscope called photomicrographs.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Organelles are the specialized sub-compartments of eukaryotic cells. Explore these infinitesimal parts of cells.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Cork (wm, 300X); Onion skin (wm, st, 200X); Green leaf (cs, st, 300X); Cheek cells (wm, st, 900X); Blood cells (sm, st, 900X); Nerve cells (cs, st, 300X); Bacteria (sm, st, 1,500X); and Virus (wm, 50,000X). Box of 15 sets also includes background subject information and reproducible student worksheets with answer keys.
Spécifications
  • Désignation:
    Microslide, Cell Structure, Set of 15 (Box)