Mirrors
Glass mirror, 4x4"
Catalog # 470020-572
Supplier: United Scientific Supplies
Supplier #: MSG4X4
Jump to:
Product Details & Documents
Traditional glass mirrors.
- Plate glass
- Silvered back
Use these top quality silvered mirrors for demonstrations and experiments in optics. Concave and convex mirrors are 50 mm dia.
Spécifications
- Désignation:Glass mirror, 4x4"