Medium Grade, 15 μm
Filter Paper, Medium Grade, 150 mm diameter
Catalog # 470019-888
Supplier: United Scientific Supplies
Fournisseur #: FPR150
Jump to:
Product Details & Documents
The most widely used qualitative filter paper for routine applications.
- Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
Packaging: Packages of 100.
Spécifications
- Désignation:Filter Paper, Medium Grade, 150 mm diameter