Cellophane
Cellophane Roll, Yellow, 20×100"
Catalog # 470019-316
Supplier: HYGLOSS PRODUCTS, INC.
Fournisseur #: 71008
Jump to:
Product Details & Documents
Use the various colors to demonstrate plants’ reaction to light waves.
- Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Spécifications
- Color:Yellow
- Désignation:Cellophane Roll
- Taille:20×100"