Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors, 140 mm (5.5"), Stainless steel, Sharp/Sharp
Catalog # 470018-940
Supplier: Dunrite Instruments
: 140981
Jump to:
Product Details & Documents
Start your cuts easily.
- Dissect without a scalpel
- Handles rounded for ambidextrous use
- Available in two sizes
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Sharp-Point Surgical Scissors
- Longueur:140 mm (5.5")
- Matière:Stainless steel
- Blade Type:Sharp/Sharp
Accessories
Informations sur le produit
- Catalog #LengthMaterialAvailabilityPrice
- Surgical Scissors