Iris Scissors
Iris Scissors, Straight, 110 mm (4.5"), Stainless steel, Sharp/Sharp
Catalog # 470018-890
Supplier: Dunrite Instruments
Fournisseur #: 140611
Jump to:
Product Details & Documents
Extra control in tight spaces.
- Fine point scissors for fine cutting
- Sharp tips give better control
- Available in straight or angled blades
Choose from straight or angled scissors, both with sharp points to facilitate finer dissection work. The angled style is often preferred for hard-to-reach tissues. First quality stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Iris Scissors, Straight
- Longueur:110 mm (4.5")
- Matière:Stainless steel
- Blade Type:Sharp/Sharp