Stake Flags | Avantor
Stake Flags
Stake Flags, Orange
Catalog # 470018-560
Supplier:   Peco Sales
Fournisseur #:  39549
Stake Flags
Catalog # 470018-560
Supplier:  Peco Sales
Stake Flags
Stake Flags
Stake Flags, Orange
Price
$38.99
Log in pour la disponibilité
Pack of 100
Product Details & Documents
A versatile marking tool.
  • Aligned with GLOBE protocols
Use these flags attached to steel wire stakes to mark areas for field or watershed studies, or use them for other activities, such as species identification. The flags are solid-colored, colorfast vinyl and flexible. They are also mold and mildew resistant. The flags are 4" x 5", the stakes are 30" high, and they come in packages of 100.
Spécifications
  • Désignation:
    Stake Flags, Orange