Increment Borers
Increment Borers
Increment Borer, 8" Bore Length
  470018-548
 :   Peco Sales
Supplier #:  63081
Increment Borers
  470018-548
Increment Borers
Increment Borers
Increment Borer, 8" Bore Length
$779.30
Each
Used to Examine Wood Quality and Measure Growth Rates
These borers are made of the finest steel and coated with Teflon to reduce friction heat, which is an important consideration when working with growing trees. The Teflon, in combination with the three-thread style, also makes the borers easy to extract.
  • Désignation:
    Increment Borer, 8" Bore Length