Utility Knife
Utility Knife, 6", 1
Catalog # 470017-314
Fournisseur #: BOS-10175
Jump to:
Discontinued
Product Details & Documents
The all-purpose knife stores five heavy-duty alloy-steel blades, included, in the handle. The blade locks in place with a safety slide.
Size: 6"L.
/!\ CAUTION: Contains functional sharp edges.
Spécifications
- Désignation:Utility Knife
- Overall Length:6"
- No. of Blades:1