Plastic Back Thermometers
Thermometer, Plastic Back, High Range, Red Alcohol Filled, −30…+110 °C (−20…+230 °F), 14 cm
Catalog # 470017-068
Supplier: PRECISION MOULDED POLYMERS LTD
Supplier #: AM1060.HB000.00000
Jump to:
Product Details & Documents
Great for Early Learning about Temperature Scales
- Plastic shell provides better protection
- Resistant to both chemical and heat
- Choose between low range or high range thermometers
Teach students about temperature readings and the difference between the Celsius and Fahrenheit with these double scale thermometers.
Spécifications
- Désignation:Thermometer, Plastic Back, High Range, Red Alcohol Filled
- Longueur:14 cm
- Range:−30…+110 °C (−20…+230 °F)