Polyethylene Disposable Gloves | Avantor
Polyethylene Disposable Gloves
Translucent, M, Box/100
Catalog # 470016-978
Fournisseur #:  GDPEMD
Polyethylene Disposable Gloves
Catalog # 470016-978
Polyethylene Disposable Gloves
Polyethylene Disposable Gloves
Translucent, M, Box/100
Price
$12.45
Log in pour la disponibilité
Pack of 50
Product Details & Documents
Ultra sensitive.
  • Ambidextrous
  • Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Spécifications
  • Taille du gant:
    M
  • Désignation:
    Translucent
  • Packaging:
    Box/100
  • Powdered/Powder-free:
    Powder-free
  • Cat. No.:
    470016-978
  • Disposable / reusable:
    Disposable
  • Ambidextrous/Hand-specific:
    Ambidextrous