Polyethylene Disposable Gloves
Translucent, M, Box/100
Catalog # 470016-978
Supplier: Dival Safety & Supplies
Fournisseur #: GDPEMD
Jump to:
Product Details & Documents
Ultra sensitive.
- Ambidextrous
- Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Spécifications
- Taille du gant:M
- Désignation:Translucent
- Packaging:Box/100
- Powdered/Powder-free:Powder-free
- Cat. No.:470016-978
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous