Solid Balls for Physics Experiments
Solid Nylon Ball, 19 mm, ¾"
Catalog # 470016-782
Supplier: UNITED STATES PLASTICS CORP.
Fournisseur #: 92062
Jump to:
Product Details & Documents
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
Spécifications
- Désignation:Solid Nylon Ball, 19 mm
- Diameter:¾"