Dropping Bottles
30 ml (1 oz.), —
Catalog # 470016-200
Supplier: Qorpak
Supplier #: 501182
Jump to:
Product Details & Documents
Dropping bottles are made of low-density polyethylene.
- White Polypropylene Dropper and Nasal Cap
- Molded Plastic Screw Cap Seals at Tip
- Low-Density Polyethylene
They feature a separable dropping tip.
The molded plastic screw cap seals at the tip.
Specifications
- Taille du bouchon:—
- Capacité:30 ml (1 oz.)