Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Spherical Mirrors | Avantor

Spherical Mirrors

Concave mirror, 150 mm, 50 mm
Catalog # 470013-306
Supplier:   Wards
Fournisseur #:  PH0520D
Spherical Mirrors
Catalog # 470013-306
Supplier:  Wards
Spherical Mirrors
Spherical Mirrors

Concave mirror, 150 mm, 50 mm

Price
$9.65
Log in pour la disponibilité
Each
Product Details & Documents

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

Spécifications
  • Désignation:
    Concave mirror
  • Focal Length:
    150 mm
  • Diameter:
    50 mm