Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Assorted Rubber Stoppers | Avantor

Assorted Rubber Stoppers

Assorted Rubber Stoppers, 1 lb., 1 to 6
Catalog # 470012-960
Assorted Rubber Stoppers
Catalog # 470012-960
Supplier:  
Assorted Rubber Stoppers
Assorted Rubber Stoppers

Assorted Rubber Stoppers, 1 lb., 1 to 6

Discontinued
Product Details & Documents

Solid, 1-hole and 2-hole stoppers in assorted sizes.

  • Superior grade of rubber
  • Soft, resilent, and durable
  • Low sulfur content
  • Size numbers molded onto the top
  • All stoppers are 25mm long
  • Aligned with GLOBE protocols

Designed to stay pliable over long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.

Caution: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.

Spécifications
  • Désignation:
    Assorted Rubber Stoppers, 1 lb.
  • Taille:
    1 to 6
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.