Wooden Splints
Wooden Splints, 114 mm, 4 mm
Catalog # 470012-768
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
SPLINT WOODEN 114X4MM PK 500 Catalog #470005-838
Product 470005-838 is a replacement for 470012-768 but varies by Brand, Packaging or Quantity.
SPLINT WOODEN 114X4MM PK 100 Catalog #470104-504
Product 470104-504 is a replacement for 470012-768 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Wooden splints are used for mixing and applying in a laboratory setting.
Spécifications
- Désignation:Wooden Splints
- Hauteur:114 mm
- Width:4 mm