Protozoa Microslide
Protozoa microslide
Catalog # 470009-144
Supplier: SUPERTEK SCIENTIFIC LLC
Fournisseur #: T-202
Jump to:
Product Details & Documents
Compare and Contrast Unicellular Organisms
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Condylostoma/Cilia and food vacuoles (200X),
Vorticella (75X), Dileptus/Trichocysts and contractile vacuoles (100X), Chlamydomonas/Flagella and chloroplasts (400X), Volvox (150X), Amoeba and Actinophrys/Pseudopods (200X), Difflugia (200X), and Radiolarians (25X).
Spécifications
- Désignation:Protozoa microslide