Basic Human Histology Microslides
Basic Human Histology Microslides, one Set
470009-118
: T-249
Provides a histological survey of body tissues
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
Contents:
Bone (cs, st, 150X)
Cartilage (69X)
Fat cells (50X)
Epithelial tissue (175X); Ciliated epithelial cell (1,400X)
Nerve cells (st, 400X)
Striated muscle cells (1,000X)
Red blood cells (em, 3,000X)
White blood cells (262X and em, 3,000X)
- Désignation:Basic Human Histology Microslides, one Set