Cell Nucleus Microslide | Avantor
Cell Nucleus Microslide
Cell nucleus microslide
Catalog # 470009-116
Fournisseur #:  T-248
Cell Nucleus Microslide
Catalog # 470009-116
Cell Nucleus Microslide
Cell Nucleus Microslide
Cell nucleus microslide
Price
19,60 $
Log in pour la disponibilité
Each
Product Details & Documents
Explore diverse cell types
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Animal cell nucleus (900X)
Plant cell nucleus (1,000X)
Nuclear reproduction (100X)
Human blood cells (900X)
Voluntary muscle cells (900X)
Nuclear membrane (em, 100,000X)
Nuclear membrane surface (em, 100,000X)
Endoplasmic reticulum (em, 125,000X)
Spécifications
  • Désignation:
    Cell nucleus microslide