Monocots and Dicots Microslide
Microslide, monocots and dicots set
Catalog # 470009-094
Supplier: SUPERTEK SCIENTIFIC LLC
Fournisseur #: T-210
Jump to:
Product Details & Documents
Flowering Plant Comparisons
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Monocot seed (ls, 20X), Dicot seed (ls, 40X), Petunia seedling (wm,10X), Monocot and dicot leaves (wm), Monocot and dicot roots (cs, 50X), Monocot and dicot stems (cs, 8X and 34X), Dicot stem; 3 yrs. old (wm,18X), and Monocot flower (cs, 6X).
Spécifications
- Désignation:Microslide, monocots and dicots set