The Chick Embryo Microslide
Product Details & Documents
Explore chick development on the microscopic level.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes 13 hours (wm, st, 1X), 18 hours (wm, st, 1X), 21 hours (wm, st, 1X), 28 hours (wm, st, 1X), 38 hours (wm, st, 1X), 48 hours(wm, st, 1X), 56 hours (wm, st,1X), and 96 hours (wm, st, 1X).
Specifications
- Désignation:The chick embryo microslide