Balls for Physics Experiments
Drilled Aluminum Ball, 19 mm, ¾"
  470007-144
 :  ALBL19
Balls for Physics Experiments
  470007-144
 :  
Balls for Physics Experiments
Balls for Physics Experiments
Drilled Aluminum Ball, 19 mm, ¾"
Ideal for Collision and Pendulum Demonstrations
Drilled balls for pendulum bobs or impact experiments. 
  • Désignation:
    Drilled Aluminum Ball, 19 mm
  • Diameter:
    ¾"