Radiation Lab Activity
Radiation Lab Activity, Materials for One Lab Group, 1
470006-730
Students investigate the absorption and radiation of energy on different surfaces.
- Activity Includes Light Source
- Two Experiments in One Activity
- Students Research Absorption and Radiation of Energy
They then turn on the light at 10 cm distance and observe how temperatures respond differently in the black can and silver can.
In the reverse experiment, the rates of cooling can be determined after students turn off the light.
: Each kit includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, and two other thermometers (-20° to 50°C). The single kit also includes one 120VAC clamp lamp with a reflector and 100 watt light bulb; the class kit includes four lamps.
- Désignation:Radiation Lab Activity, Materials for One Lab Group
- No. of Lab Groups:1