Bare Copper Wire, 113 g Spool | Avantor
Bare Copper Wire, 113 g Spool
Wire, Bare Copper, 30 Gauge, 259 m, 113 grams
Catalog # 470005-942
Supplier:   Arcor Electronics
Fournisseur #:  B30
Bare Copper Wire, 113 g Spool
Catalog # 470005-942
Supplier:  Arcor Electronics
Bare Copper Wire, 113 g Spool
Bare Copper Wire, 113 g Spool
Wire, Bare Copper, 30 Gauge, 259 m, 113 grams
Liquidation
9,55 $
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A bare copper wire of the highest quality. Available in several standard AWG sizes.
  • 113 g spool
Spécifications
  • Désignation:
    Wire, Bare Copper, 30 Gauge, 259 m, 113 grams