Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Fine-Point Forceps With Guide Pins | Avantor

Fine-Point Forceps With Guide Pins

Fine-Point Forceps With Guide Pin, Stainless steel, 115 mm (4.5"), Curved
Catalog # 470005-440
Supplier:   Dunrite Instruments
Fournisseur #:  140531
Fine-Point Forceps With Guide Pins
Catalog # 470005-440
Fine-Point Forceps With Guide Pins
Fine-Point Forceps With Guide Pins

Fine-Point Forceps With Guide Pin, Stainless steel, 115 mm (4.5"), Curved

Price
$7.05
Log in pour la disponibilité
Each Box of 50
Product Details & Documents

Slip proof fine point forceps.

  • Guide pins keep tip aligned
  • Prevents slipping and twisting when closed
  • Textured tips improve hold on slippery material

Used in our own labs, these forceps feature a built-in guide pin to keep the fine tips aligned, which is important for grasping individual vessels or nerves. Stainless steel. Length: 4¹/₂".

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Spécifications
  • Désignation:
    Fine-Point Forceps With Guide Pin
  • Longueur:
    115 mm (4.5")
  • Matière:
    Stainless steel
  • Tip Style:
    Curved