Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Pulley Cords | Avantor

Pulley Cords

Braided Nylon pulley cord, tensile strength 200 lbs. test, White, 9 m (10 yds.)
Catalog # 470005-414
Supplier:   Arcor Electronics
Fournisseur #:  SDX16165
Pulley Cords
Catalog # 470005-414
Supplier:  Arcor Electronics
Pulley Cords
Pulley Cords

Braided Nylon pulley cord, tensile strength 200 lbs. test, White, 9 m (10 yds.)

Price
$31.00
Log in pour la disponibilité
Each
Product Details & Documents

Ideal for small pulley systems, cord is available in a variety of colors, strengths, and lengths.

  • Ideal for pulley systems, hanging masses, and wave demonstrations
  • Nylon and Dacron are extremely durable
  • Ends may be heat fused to prevent fraying
  • Premium Dacron cords resists stretching and fraying

Every physics classroom needs good-quality string - whether it's for an Atwood's machine, pulling friction blocks, or hanging pendulums. Teachers need string that is easy to knot and won't unravel after it is cut. These products have been selected for durability and strength, and come on a convenient spool for easy dispensing and storage.

Spécifications
  • Color:
    White
  • Désignation:
    Braided Nylon pulley cord, tensile strength 200 lbs. test
  • Longueur:
    9 m (10 yds.)
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Support base steel for braided nylon pulley cord, tensile strength 15 lbs. test, height: 23 mm, base dimension: 5×7", thread size: ³/₈-16
    Support base steel for braided nylon pulley cord, tensile strength 15 lbs. test, height: 23 mm, base dimension: 5×7", thread size: ³/₈-16
    470123-472
    Log in pour la disponibilité
    Each - $36.65

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.