Polypropylene Beaker Set
50 ml, 100 ml, 250 ml, 500 ml, 1000 ml, Set of 5
Catalog # 470005-162
Supplier: Dynalon
: 222195
Jump to:
Product Details & Documents
Unbreakable polypropylene.
- Autoclavable
- Chemical-resistant
These beaker sets autoclavable up to 145 °C and chemical resistant. All feature pour spouts and metric graduation scales.
Spécifications
- Capacité:50 ml, 100 ml, 250 ml, 500 ml, 1000 ml
- Désignation:Set of 5